Assay Details
Target Gene Details
Entrez Gene ID: | 218624 |
Gene Name: | interleukin 31 receptor A |
Gene Aliases: |
CRL3, GLM-R, GPL, Glmr, NR10 |
Location: |
Chr.13:112521849-112591399 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il31ra | NM_139299.2 | 14 | 1864 | NP_647460.2 |
AB083111.1 | 14 | 1863 | BAB88745.1 | |
AF486621.1 | 11 | 1443 | AAM27959.1 | |
AK030512.1 | 3 | 433 | BAC26998.1 | |
BC066164.1 | AAH66164.1 |
More Information
Set Membership: |
Intragenic Exonic |