Assay Details
Target Gene Details
Entrez Gene ID: | 76487 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 3G |
Gene Aliases: |
1600032L17Rik |
Location: |
Chr.13:35958799-35970388 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r3g |