Assay Details
Target Gene Details
Entrez Gene ID: | 66336 |
Gene Name: | centromere protein P |
Gene Aliases: |
1700022C02Rik, 4921518G09Rik |
Location: |
Chr.13:49464025-49652879 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cenpp | NM_025495.3 | NP_079771.1 | ||
AK006234.1 | BAB24472.1 | |||
AK012863.1 | BAB28519.1 | |||
AK014919.1 | BAB29620.1 | |||
AK076577.1 | BAC36401.1 | |||
BC147207.1 | ||||
BC147208.1 |
Target Gene Details
Entrez Gene ID: | 18295 |
Gene Name: | osteoglycin |
Gene Aliases: |
3110079A16Rik, OG, OIF, SLRR3A, mimecan, mimican |
Location: |
Chr.13:49608071-49624500 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ogn | NM_008760.4 | 4 | 511 | NP_032786.1 |
AK028300.1 | 4 | 537 | ||
AK053650.1 | 4 | 512 | BAC35462.1 | |
AK075638.1 | 3 | 468 | ||
AK160760.1 | 4 | 533 | BAE35995.1 | |
AK163772.1 | 3 | 464 | ||
BC021939.1 | 3 | 414 | AAH21939.1 | |
D31951.1 | 3 | 424 | BAA06721.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |