Assay Details
Target Gene Details
Entrez Gene ID: | 407785 |
Gene Name: | NADH dehydrogenase (ubiquinone) Fe-S protein 6 |
Gene Aliases: |
BC059730, IP13 |
Location: |
Chr.13:73319876-73328482 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufs6 | NM_010888.2 | NP_035018.1 | ||
NR_033306.1 | 1 | 172 | ||
AK147147.1 | 1 | 141 | ||
BC086933.1 | AAH86933.1 | |||
BG081040.2 |
More Information
Set Membership: |
Intragenic Exonic |