Assay Details
Target Gene Details
Entrez Gene ID: | 319554 |
Gene Name: | isopentenyl-diphosphate delta isomerase |
Gene Aliases: |
4832416K17Rik |
Location: |
Chr.13:8885606-8892396 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Idi1 | NM_145360.2 | NP_663335.2 | ||
AK029302.1 | BAC26382.1 | |||
AK084208.1 | ||||
AK132073.1 | ||||
AK160832.1 | BAE36037.1 | |||
BC004801.1 | AAH04801.1 | |||
BC110313.1 | AAI10314.1 | |||
CT010310.1 | CAJ18518.1 |
More Information
Set Membership: |
Intragenic Non-exonic |