Assay Details
Target Gene Details
Entrez Gene ID: | 56541 |
Gene Name: | hyaluronic acid binding protein 4 |
Gene Aliases: |
4933413D03Rik, 4933428J01Rik |
Location: |
Chr.13:64161866-64186537 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Habp4 | NM_019986.3 | NP_064370.2 | ||
AF227684.1 | AAF36966.1 | |||
AK016800.1 | BAB30437.1 | |||
AK144206.1 | BAE25769.1 | |||
AK146525.1 | BAE27234.1 | |||
BC148304.2 |
More Information
Set Membership: |
Intragenic Non-exonic |