Assay Details
Target Gene Details
Entrez Gene ID: | 73274 |
Gene Name: | GC-rich promoter binding protein 1 |
Gene Aliases: |
1700034P14Rik, AU019836, D230035M11Rik, Gpbp, mGPBP |
Location: |
Chr.13:111425680-111490123 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpbp1 | NM_001122963.1 | NP_001116435.1 | ||
NM_028487.4 | NP_082763.3 | |||
AK135728.1 | BAE22631.1 | |||
AK136894.1 | ||||
AK145318.1 | BAE26364.1 | |||
AK151283.1 | ||||
AK163370.1 | BAE37320.1 | |||
AK163390.1 | BAE37330.1 | |||
AK169406.1 | BAE41152.1 | |||
AY382529.1 | AAQ88446.1 | |||
BC016083.1 | AAH16083.1 | |||
BC067075.1 | AAH67075.1 | |||
BC094888.1 | AAH94888.1 |