Assay Details
Target Gene Details
Entrez Gene ID: | 18260 |
Gene Name: | occludin |
Gene Aliases: |
AI503564, Ocl |
Location: |
Chr.13:100496504-100553689 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ocln | NM_008756.2 | 6 | 1362 | NP_032782.1 |
AK019880.1 | 6 | 1314 | BAB31900.1 | |
AK033532.1 | 6 | 1295 | ||
AK042576.1 | 6 | 1362 | BAC31298.1 | |
AK145798.1 | 4 | 521 | BAE26656.1 | |
BC138679.1 | 6 | 1345 | ||
BC138680.1 | 6 | 1345 | ||
BC145371.1 | 6 | 1360 | ||
U49185.1 | 6 | 1405 | AAC52515.1 |
More Information
Set Membership: |
Intragenic Exonic |