Assay Details
Target Gene Details
Entrez Gene ID: | 14945 |
Gene Name: | granzyme K |
Gene Aliases: |
- |
Location: |
Chr.13:113171874-113180897 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gzmk | NM_008196.1 | 3 | 282 | NP_032222.1 |
BC117981.1 | 3 | 281 |
More Information
Set Membership: |
Intragenic Exonic |