Assay Details
Target Gene Details
Entrez Gene ID: | 218121 |
Gene Name: | membrane bound O-acyltransferase domain containing 1 |
Gene Aliases: |
9130215M02Rik, BC023845, LPEAT1, Moact1, Oact1 |
Location: |
Chr.13:30136449-30246694 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mboat1 | NM_153546.4 | NP_705774.1 | ||
AB297382.1 | ||||
AB305045.1 | ||||
AK033388.1 | BAC28261.1 | |||
AK137755.1 | ||||
AK146984.1 | BAE27587.1 | |||
BC023845.1 | AAH23845.1 | |||
BC024653.1 | AAH24653.1 |
More Information
Set Membership: |
Intragenic Non-exonic |