Assay Details
Target Gene Details
Entrez Gene ID: | 28114 |
Gene Name: | NOL1/NOP2/Sun domain family member 2 |
Gene Aliases: |
D13Wsu123e, Misu |
Location: |
Chr.13:69612016-69635780 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nsun2 | NM_145354.5 | NP_663329.3 | ||
AK030124.1 | BAC26795.1 | |||
AK075999.1 | BAC36110.1 | |||
AK150631.1 | BAE29720.1 | |||
AK151917.1 | BAE30795.1 | |||
AK154789.1 | ||||
AK171399.1 | ||||
BC013625.1 | AAH13625.1 | |||
DQ490066.1 |
More Information
Set Membership: |
Intragenic Non-exonic |