Assay Details
Target Gene Details
Entrez Gene ID: | 15357 |
Gene Name: | 3-hydroxy-3-methylglutaryl-Coenzyme A reductase |
Gene Aliases: |
HMG-CoAR, Red |
Location: |
Chr.13:96648962-96670936 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hmgcr | NM_008255.2 | 19 | 2647 | NP_032281.2 |
AK079302.1 | 7 | 846 | BAC37602.1 | |
AK159899.1 | 18 | 2535 | ||
AK198607.1 | 3 | 262 | ||
BC059873.1 | 10 | 1328 | AAH59873.1 | |
BC085083.1 | 19 | 2559 | AAH85083.1 | |
M62766.1 | 4 | 509 | AAA37819.1 |
More Information
Set Membership: |
Intragenic Exonic |