Assay Details
Target Gene Details
Entrez Gene ID: | 59079 |
Gene Name: | Erbb2 interacting protein |
Gene Aliases: |
1700028E05Rik, Erbin, mKIAA1225 |
Location: |
Chr.13:103818786-103920586 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Erbb2ip | NM_001005868.2 | NP_001005868.1 | ||
NM_001289473.1 | NP_001276402.1 | |||
NM_001289474.1 | NP_001276403.1 | |||
NM_001289475.1 | NP_001276404.1 | |||
AF263743.1 | AAF77047.1 | |||
AK029054.1 | BAC26267.1 | |||
AK046880.1 | 1 | 2089 | ||
AK051733.1 | BAC34742.1 | |||
AK122473.1 | BAC65755.1 | |||
BC005691.1 | ||||
BC028256.1 | AAH28256.1 | |||
BC141178.1 | ||||
BC141188.1 | ||||
BC145498.1 | ||||
BC145509.1 | ||||
EU233994.1 |
More Information
Set Membership: |
Intragenic Non-exonic |