Assay Details
Target Gene Details
Entrez Gene ID: | 59079 |
Gene Name: | Erbb2 interacting protein |
Gene Aliases: |
1700028E05Rik, Erbin, mKIAA1225 |
Location: |
Chr.13:103818786-103920586 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Erbb2ip | NM_001005868.2 | 16 | 1763 | NP_001005868.1 |
NM_001289473.1 | 16 | 1763 | NP_001276402.1 | |
NM_001289474.1 | 16 | 1763 | NP_001276403.1 | |
NM_001289475.1 | 16 | 1763 | NP_001276404.1 | |
AK029054.1 | 5 | 445 | BAC26267.1 | |
AK122473.1 | 16 | 1442 | BAC65755.1 | |
AK169529.1 | 16 | 1692 | ||
BC141178.1 | 16 | 1472 | ||
BC141188.1 | 16 | 1472 | ||
BC145498.1 | 16 | 1472 | ||
BC145509.1 | 16 | 1472 | ||
EU233994.1 | 18 | 2520 |
More Information
Set Membership: |
Intragenic Exonic |