Assay Details
Target Gene Details
Entrez Gene ID: | 15357 |
Gene Name: | 3-hydroxy-3-methylglutaryl-Coenzyme A reductase |
Gene Aliases: |
HMG-CoAR, Red |
Location: |
Chr.13:96648962-96670936 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hmgcr | NM_008255.2 | NP_032281.2 | ||
AK079302.1 | BAC37602.1 | |||
AK159899.1 | ||||
AK198607.1 | ||||
BC059873.1 | AAH59873.1 | |||
BC085083.1 | AAH85083.1 | |||
M62766.1 | AAA37819.1 |
More Information
Set Membership: |
Intragenic Non-exonic |