Assay Details
Target Gene Details
Entrez Gene ID: | 15357 |
Gene Name: | 3-hydroxy-3-methylglutaryl-Coenzyme A reductase |
Gene Aliases: |
HMG-CoAR, Red |
Location: |
Chr.13:96648962-96670936 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hmgcr | NM_008255.2 | 12 | 1612 | NP_032281.2 |
AK159899.1 | 11 | 1500 | ||
BC059873.1 | 3 | 293 | AAH59873.1 | |
BC085083.1 | 12 | 1524 | AAH85083.1 |
More Information
Set Membership: |
Intragenic Exonic |