Assay Details
Target Gene Details
Entrez Gene ID: | 238871 |
Gene Name: | phosphodiesterase 4D, cAMP specific |
Gene Aliases: |
9630011N22Rik, Dpde3 |
Location: |
Chr.13:108654177-109955969 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde4d | NM_011056.3 | 3 | 379 | NP_035186.1 |
AF536978.1 | 3 | 379 | AAN10120.1 | |
BC132107.1 | 2 | 247 | ||
BC132109.1 | 2 | 247 |
More Information
Set Membership: |
Intragenic Exonic |