Assay Details
Target Gene Details
Entrez Gene ID: | 238871 |
Gene Name: | phosphodiesterase 4D, cAMP specific |
Gene Aliases: |
9630011N22Rik, Dpde3 |
Location: |
Chr.13:108654177-109955969 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde4d | NM_011056.3 | 10 | 1093 | NP_035186.1 |
AF536978.1 | 10 | 1093 | AAN10120.1 | |
AK081466.1 | 3 | 466 | BAC38226.1 | |
AY388962.1 | 8 | 706 | AAQ90406.1 | |
BC132107.1 | 9 | 961 | ||
BC132109.1 | 9 | 961 | ||
DQ665891.1 | 3 | 563 | ||
DQ665892.1 | 3 | 349 | ||
DQ665893.1 | 4 | 477 | ||
DQ665894.1 | 4 | 321 | ||
DQ665895.1 | 3 | 604 | ||
DQ665896.1 | 3 | 393 | ||
EU489880.1 | 8 | 949 |
More Information
Set Membership: |
Intragenic Exonic |