Assay Details
Target Gene Details
Entrez Gene ID: | 17152 |
Gene Name: | male germ cell-associated kinase |
Gene Aliases: |
A930010O05Rik |
Location: |
Chr.13:41025007-41079706 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mak | NM_001145802.1 | NP_001139274.1 | ||
NM_001145803.1 | NP_001139275.1 | |||
NM_008547.2 | NP_032573.2 | |||
BQ930470.1 | ||||
X66983.1 | CAA47392.1 |
More Information
Set Membership: |
Intragenic Non-exonic |