Assay Details
Target Gene Details
Entrez Gene ID: | 14780 |
Gene Name: | glutathione peroxidase 5 |
Gene Aliases: |
AV379049, Arep |
Location: |
Chr.13:21286429-21292729 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpx5 | NM_010343.2 | NP_034473.2 | ||
AK029731.1 | BAC26586.1 | |||
BC100749.1 | AAI00750.1 | |||
BC103588.1 | AAI03589.1 | |||
BC103589.1 | AAI03590.1 | |||
BC105648.1 | ||||
X53780.1 | CAA37796.1 |
More Information
Set Membership: |
Intragenic Non-exonic |