Assay Details
Target Gene Details
Entrez Gene ID: | 80898 |
Gene Name: | endoplasmic reticulum aminopeptidase 1 |
Gene Aliases: |
Arts1, ERAAP, PILSA, PILSAP |
Location: |
Chr.13:74639624-74691931 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Erap1 | NM_030711.4 | 17 | 2560 | NP_109636.1 |
AB047552.1 | 16 | 2548 | BAB11982.1 | |
AF227511.1 | 16 | 2536 | AAG44260.1 | |
AK029841.1 | 17 | 2559 | ||
AK030329.1 | 17 | 2561 | BAC26904.1 | |
AK040350.1 | 14 | 1778 | BAC30569.1 | |
AK159406.1 | 17 | 2561 | ||
BC046610.1 | 17 | 2566 | AAH46610.1 |
More Information
Set Membership: |
Intragenic Exonic |