Assay Details
Target Gene Details
Entrez Gene ID: | 320806 |
Gene Name: | G elongation factor, mitochondrial 2 |
Gene Aliases: |
6530419G12Rik, A930009M04Rik, EF-G2mt, EFG2, MST027, RRF2mt |
Location: |
Chr.13:97137937-97181196 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gfm2 | NM_001146043.2 | 18 | 1883 | NP_001139515.1 |
NM_001271463.1 | 18 | 1883 | NP_001258392.1 | |
NM_001271464.1 | 17 | 1802 | NP_001258393.1 | |
NM_001271465.1 | 18 | 1910 | NP_001258394.1 | |
NM_177266.5 | 18 | 1916 | NP_796240.3 | |
AK035819.1 | 18 | 1821 | ||
AK044371.1 | 17 | 1803 | BAC31889.1 | |
AK052633.1 | 2 | 226 | ||
AK083441.1 | 17 | 1922 | ||
AK161797.1 | 18 | 1819 | BAE36579.1 | |
AK165871.1 | 18 | 1877 | BAE38428.1 | |
BC027341.1 | 18 | 1867 | AAH27341.1 | |
BC138233.1 | 18 | 1814 | ||
BC145208.1 | 18 | 1814 |
More Information
Set Membership: |
Intragenic Exonic |