Assay Details
Target Gene Details
Entrez Gene ID: | 15212 |
Gene Name: | hexosaminidase B |
Gene Aliases: |
- |
Location: |
Chr.13:97176331-97198357 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hexb | NM_010422.2 | NP_034552.1 | ||
AK151149.1 | BAE30155.1 | |||
AK159134.1 | BAE34846.1 | |||
AK159178.1 | BAE34877.1 | |||
AK160560.1 | BAE35874.1 | |||
AK168825.1 | BAE40652.1 | |||
AK209131.1 | ||||
U07633.1 | AAA18776.1 | |||
Y00964.1 | CAA68781.1 |
More Information
Set Membership: |
Intragenic Non-exonic |