Assay Details
Target Gene Details
Entrez Gene ID: | 75678 |
Gene Name: | inositol 1,3,4,5,6-pentakisphosphate 2-kinase |
Gene Aliases: |
1810043M15Rik, InsP6 |
Location: |
Chr.13:49421223-49463060 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ippk | NM_001276399.1 | NP_001263328.1 | ||
NM_199056.3 | NP_951011.1 | |||
NR_075085.1 | ||||
AK029085.1 | BAE20447.1 | |||
AK031987.1 | ||||
AK160092.1 | BAE35623.1 | |||
AK167467.1 | BAE39551.1 | |||
BC024630.1 | ||||
BC062167.1 | AAH62167.1 | |||
BC065157.1 | AAH65157.1 | |||
DV647168.1 |
More Information
Set Membership: |
Intragenic Non-exonic |