Assay Details
Target Gene Details
Entrez Gene ID: | 320806 |
Gene Name: | G elongation factor, mitochondrial 2 |
Gene Aliases: |
6530419G12Rik, A930009M04Rik, EF-G2mt, EFG2, MST027, RRF2mt |
Location: |
Chr.13:97137937-97181196 on Build GRCm38 |
Assay Gene Location: | Within Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gfm2 | NM_001146043.2 | NP_001139515.1 | ||
AK044750.1 | 1 | 1725 | ||
AK161797.1 | BAE36579.1 |
Target Gene Details
Entrez Gene ID: | 15212 |
Gene Name: | hexosaminidase B |
Gene Aliases: |
- |
Location: |
Chr.13:97176331-97198357 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hexb | NM_010422.2 | 11 | 1368 | NP_034552.1 |
AK151149.1 | 11 | 1305 | BAE30155.1 | |
AK159134.1 | 11 | 1321 | BAE34846.1 | |
AK159178.1 | 11 | 1342 | BAE34877.1 | |
AK160560.1 | 11 | 1337 | BAE35874.1 | |
AK168825.1 | 11 | 1342 | BAE40652.1 | |
CD775639.1 | 4 | 555 | ||
U07633.1 | 11 | 1340 | AAA18776.1 | |
Y00964.1 | 11 | 1281 | CAA68781.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |