Assay Details
Target Gene Details
Entrez Gene ID: | 94253 |
Gene Name: | HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1 |
Gene Aliases: |
9330116H24Rik, AV273951, E130207I19Rik, Nedl1 |
Location: |
Chr.13:14226438-14523280 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hecw1 | NM_001081348.3 | 28 | 5041 | NP_001074817.3 |
AB083710.1 | 26 | 4544 | BAB97389.1 | |
AK033922.1 | 8 | 976 | BAC28516.1 | |
AK172932.1 | 19 | 3326 | BAD32210.1 |
More Information
Set Membership: |
Intragenic Exonic |