Assay Details
Target Gene Details
Entrez Gene ID: | 66254 |
Gene Name: | DIM1 dimethyladenosine transferase 1-like (S. cerevisiae) |
Gene Aliases: |
1500031M22Rik, AV111347, Dimt1l |
Location: |
Chr.13:106947043-106960225 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dimt1 | NM_025447.4 | NP_079723.1 | ||
AK007759.1 | ||||
AK011544.1 | BAB27687.1 | |||
AK015145.1 | ||||
AK078186.1 | BAC37166.1 | |||
AK078401.1 | ||||
AK146188.1 | BAE26964.1 | |||
AK161333.1 | BAE36330.1 | |||
BC019799.1 | AAH19799.1 | |||
BF608861.1 |
More Information
Set Membership: |
Intragenic Non-exonic |