Assay Details
Target Gene Details
Entrez Gene ID: | 107889 |
Gene Name: | glial cells missing homolog 2 (Drosophila) |
Gene Aliases: |
Gcm-rs1, Gcm1, Gcm1-rs2 |
Location: |
Chr.13:41101427-41111035 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gcm2 | NM_008104.2 | NP_032130.2 | ||
AF081556.1 | AAC64782.1 | |||
AK132499.1 | ||||
AK135689.1 | ||||
BC110631.1 | AAI10632.1 | |||
BC110632.1 | AAI10633.1 | |||
D88611.1 | BAA13649.1 |