Assay Details
Target Gene Details
Entrez Gene ID: | 218236 |
Gene Name: | family with sequence similarity 120, member A |
Gene Aliases: |
C9orf10, Ossa |
Location: |
Chr.13:48879216-48967877 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam120a | NM_001033268.2 | 2 | 598 | NP_001028440.2 |
AK172909.1 | 2 | 787 | BAD32187.1 | |
BC042582.1 | 1 | 114 | ||
BC158134.1 | 2 | 590 |