Assay Details
Target Gene Details
Entrez Gene ID: | 94066 |
Gene Name: | mitochondrial ribosomal protein L36 |
Gene Aliases: |
AI646041 |
Location: |
Chr.13:73331009-73332178 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mrpl36 | NM_053163.1 | NP_444393.1 | ||
AB049655.1 | BAB40860.1 | |||
AK075780.1 | BAC35954.1 | |||
AK131597.1 | BAE20708.1 | |||
AK131601.1 | BAE20711.1 | |||
AK131604.1 | BAE20714.1 | |||
AK160778.1 | BAE36002.1 | |||
BC009166.1 | AAH09166.1 |
More Information
Set Membership: |
Intragenic Non-exonic |