Assay Details
Target Gene Details
Entrez Gene ID: | 17434 |
Gene Name: | molybdenum cofactor synthesis 2 |
Gene Aliases: |
AI415403 |
Location: |
Chr.13:114818237-114829420 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mocs2 | NM_001113374.1 | NP_001106845.1 | ||
NM_001113375.1 | NP_001106846.1 | |||
NM_013826.3 | NP_038854.2 | |||
AF091872.1 | AAD14600.1 | |||
AK002719.1 | BAB22306.1 | |||
AK010112.1 | BAB26707.2 | |||
AK078782.1 | BAC37393.1 | |||
BC024371.1 | AAH24371.1 | |||
CX220128.1 |
More Information
Set Membership: |
Intragenic Non-exonic |