Assay Details
Target Gene Details
Entrez Gene ID: | 14186 |
Gene Name: | fibroblast growth factor receptor 4 |
Gene Aliases: |
Fgfr-4 |
Location: |
Chr.13:55152818-55168759 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fgfr4 | NM_008011.2 | NP_032037.2 | ||
AK084850.1 | BAC39292.1 | |||
AK085723.1 | ||||
AY493377.2 | AAS72387.2 | |||
BC033313.1 | AAH33313.1 | |||
DQ388428.1 | ABD43187.1 | |||
X59927.1 | CAA42551.1 |
More Information
Set Membership: |
Intragenic Non-exonic |