Assay Details
Target Gene Details
Entrez Gene ID: | 67241 |
Gene Name: | structural maintenance of chromosomes 6 |
Gene Aliases: |
2810489L22Rik, 3830418C19Rik, AA990493, AU018782, AW742439, SMC-6, Smc6l1, mKIAA4103 |
Location: |
Chr.12:11265886-11319785 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smc6 | NM_025695.4 | NP_079971.2 | ||
AJ310552.1 | CAC39250.1 | |||
AK009009.1 | BAB26022.3 | |||
AK011982.1 | BAB27956.1 | |||
AK012626.1 | BAB28365.1 | |||
AK019823.1 | ||||
AK050341.1 | BAC34200.2 | |||
AK088002.1 | BAC40087.2 | |||
AK135873.1 | BAE22709.1 | |||
AK148478.1 | BAE28576.1 | |||
AK220488.1 | BAD90287.1 | |||
BC048790.1 | AAH48790.1 | |||
BC090630.1 | AAH90630.1 | |||
BC099955.1 | AAH99955.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |