Assay Details
Target Gene Details
Entrez Gene ID: | 67241 |
Gene Name: | structural maintenance of chromosomes 6 |
Gene Aliases: |
2810489L22Rik, 3830418C19Rik, AA990493, AU018782, AW742439, SMC-6, Smc6l1, mKIAA4103 |
Location: |
Chr.12:11265886-11319785 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smc6 | NM_025695.4 | 14 | 1906 | NP_079971.2 |
AJ310552.1 | 13 | 1498 | CAC39250.1 | |
AK084304.1 | 1 | 79 | ||
AK135873.1 | 14 | 1644 | BAE22709.1 | |
AK148478.1 | 14 | 1671 | BAE28576.1 | |
AK220488.1 | 13 | 1581 | BAD90287.1 | |
BC090630.1 | 14 | 1906 | AAH90630.1 | |
BC099955.1 | 14 | 1593 | AAH99955.1 |
More Information
Set Membership: |
Intragenic Exonic |