Assay Details
Target Gene Details
Entrez Gene ID: | 69675 |
Gene Name: | peroxidasin homolog (Drosophila) |
Gene Aliases: |
2310075M15Rik, C85409, E330004E07, VPO1, mKIAA0230 |
Location: |
Chr.12:29937877-30017658 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pxdn | NM_181395.2 | NP_852060.2 | ||
AK088837.1 | ||||
AK122223.1 | BAC65505.1 | |||
AK142872.1 | BAE25216.1 | |||
BC112913.1 | ||||
BC150789.1 |
More Information
Set Membership: |
Intragenic Non-exonic |