Assay Details
Target Gene Details
Entrez Gene ID: | 544971 |
Gene Name: | B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB |
Gene Aliases: |
AI662220, AW049390, AW456745, B130055N23Rik, G630013P12Rik, TAF3B1, TFC5, TFIIIB150, TFIIIB90, Tfnr, mKIAA1241 |
Location: |
Chr.13:100017994-100104073 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 29 - Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bdp1 | NM_001081061.1 | NP_001074530.1 | ||
AK220262.1 | BAD90187.1 | |||
BC030359.1 | AAH30359.1 | |||
BC144852.1 |
More Information
Set Membership: |
Intragenic Non-exonic |