Assay Details
Target Gene Details
Entrez Gene ID: | 544971 |
Gene Name: | B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB |
Gene Aliases: |
AI662220, AW049390, AW456745, B130055N23Rik, G630013P12Rik, TAF3B1, TFC5, TFIIIB150, TFIIIB90, Tfnr, mKIAA1241 |
Location: |
Chr.13:100017994-100104073 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bdp1 | NM_001081061.1 | 22 | 4678 | NP_001074530.1 |
AK220262.1 | 12 | 2926 | BAD90187.1 | |
BC144852.1 | 3 | 368 |
More Information
Set Membership: |
Intragenic Exonic |