Assay Details
Target Gene Details
Entrez Gene ID: | 73086 |
Gene Name: | ribosomal protein S6 kinase, polypeptide 5 |
Gene Aliases: |
3110005L17Rik, 6330404E13Rik, AI854034, MSK1, MSPK1, RLPK |
Location: |
Chr.12:100548433-100725382 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rps6ka5 | NM_153587.2 | 13 | 1722 | NP_705815.1 |
AK030051.1 | ||||
AK032316.1 | BAC27809.1 | |||
AK046319.1 | ||||
AK160758.1 | BAE35993.1 | |||
AY341873.1 | 14 | 1711 | AAQ24158.1 | |
AY341881.1 | AAQ24165.1 | |||
BC035298.1 | 14 | 1722 | AAH35298.1 |
More Information
Set Membership: |
Intragenic Exonic |