Assay Details
Target Gene Details
Entrez Gene ID: | 69675 |
Gene Name: | peroxidasin homolog (Drosophila) |
Gene Aliases: |
2310075M15Rik, C85409, E330004E07, VPO1, mKIAA0230 |
Location: |
Chr.12:29937877-30017658 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pxdn | NM_181395.2 | 11 | 1431 | NP_852060.2 |
AK122223.1 | 11 | 1201 | BAC65505.1 | |
AK142872.1 | 11 | 1431 | BAE25216.1 | |
BC112913.1 | 2 | 228 | ||
BC150789.1 | 11 | 1426 |
More Information
Set Membership: |
Intragenic Exonic |