Assay Details
Target Gene Details
Entrez Gene ID: | 74335 |
Gene Name: | X-ray repair complementing defective repair in Chinese hamster cells 3 |
Gene Aliases: |
4432412E01Rik, AI182522, AW537713 |
Location: |
Chr.12:111803192-111813841 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Xrcc3 | NM_028875.2 | 4 | 428 | NP_083151.1 |
AK014491.1 | 4 | 428 | BAB29391.1 | |
BC043073.1 | 3 | 393 | AAH43073.1 |
More Information
Set Membership: |
Intragenic Exonic |