Assay Details
Target Gene Details
Entrez Gene ID: | 74413 |
Gene Name: | tandem C2 domains, nuclear |
Gene Aliases: |
4933406D09Rik, Mtac2d1, Tac2-N |
Location: |
Chr.12:101645446-101719000 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tc2n | NM_001082976.2 | 9 | 1118 | NP_001076445.1 |
NM_001286364.1 | 9 | 1781 | NP_001273293.1 | |
NM_028924.3 | 9 | 1470 | NP_083200.1 | |
AB062282.1 | 8 | 950 | BAB62893.1 | |
AK016683.1 | 9 | 1802 | BAB30378.1 | |
AK136558.1 | 9 | 1119 | BAE23043.1 | |
BC120793.1 | 9 | 1470 |