Assay Details
Target Gene Details
Entrez Gene ID: | 20741 |
Gene Name: | spectrin beta, erythrocytic |
Gene Aliases: |
AI842465, D330027P03Rik, Gm1301, Spnb-1, Spnb1, ja, jaundiced, mKIAA4219 |
Location: |
Chr.12:76580488-76710547 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 31 - Exon 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sptb | NM_013675.3 | NP_038703.3 | ||
AK147299.1 | BAE27831.1 | |||
AK147684.1 | BAE28073.1 | |||
AK147704.1 | BAE28085.1 | |||
AK220551.1 | BAD90537.1 | |||
BC117711.2 | ||||
BC129806.1 | ||||
BC150783.1 | ||||
S66283.1 | AAB28600.1 |