Assay Details
Target Gene Details
Entrez Gene ID: | 20741 |
Gene Name: | spectrin beta, erythrocytic |
Gene Aliases: |
AI842465, D330027P03Rik, Gm1301, Spnb-1, Spnb1, ja, jaundiced, mKIAA4219 |
Location: |
Chr.12:76580488-76710547 on Build GRCm38 |
Assay Gene Location: | Within Exon 30 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sptb | NM_013675.3 | 30 | 6793 | NP_038703.3 |
AK147299.1 | 30 | 6794 | BAE27831.1 | |
AK147684.1 | 30 | 6386 | BAE28073.1 | |
AK147704.1 | 30 | 6386 | BAE28085.1 | |
AK220551.1 | 11 | 1893 | BAD90537.1 | |
BC117711.2 | 13 | 2047 | ||
BC129806.1 | 29 | 6126 | ||
BC150783.1 | 29 | 6159 | ||
S66283.1 | 30 | 6269 | AAB28600.1 |