Assay Details
Target Gene Details
Entrez Gene ID: | 66761 |
Gene Name: | RIKEN cDNA 4933417A18 gene |
Gene Aliases: |
- |
Location: |
Chr.13:34924405-34956218 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4933417A18Rik | NM_025750.3 | 3 | 615 | NP_080026.2 |
AK016778.1 | 3 | 580 | BAB30424.1 | |
AK016838.1 | 3 | 616 | BAB30456.1 | |
BC100450.1 | 3 | 605 | AAI00451.1 | |
BC145747.1 | 3 | 570 |