Assay Details
Target Gene Details
Entrez Gene ID: | 22018 |
Gene Name: | thyroid peroxidase |
Gene Aliases: |
- |
Location: |
Chr.12:30054661-30132984 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tpo | NM_009417.2 | NP_033443.1 | ||
AK047843.1 | BAC33171.1 | |||
BC099421.1 | AAH99421.1 | |||
X60703.1 | CAA43114.1 |
More Information
Set Membership: |
Intragenic Non-exonic |