Assay Details
Target Gene Details
Entrez Gene ID: | 74413 |
Gene Name: | tandem C2 domains, nuclear |
Gene Aliases: |
4933406D09Rik, Mtac2d1, Tac2-N |
Location: |
Chr.12:101645446-101719000 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tc2n | NM_001082976.2 | NP_001076445.1 | ||
AK136558.1 | BAE23043.1 |