Assay Details
Target Gene Details
Entrez Gene ID: | 22018 |
Gene Name: | thyroid peroxidase |
Gene Aliases: |
- |
Location: |
Chr.12:30054661-30132984 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tpo | NM_009417.2 | 10 | 1765 | NP_033443.1 |
AK047843.1 | 10 | 1744 | BAC33171.1 | |
X60703.1 | 10 | 1765 | CAA43114.1 |
More Information
Set Membership: |
Intragenic Exonic |