Assay Details
Target Gene Details
Entrez Gene ID: | 20688 |
Gene Name: | trans-acting transcription factor 4 |
Gene Aliases: |
5730497N03Rik, HF-1b, HF1-b |
Location: |
Chr.12:118231686-118301440 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sp4 | NM_001166385.2 | 5 | 2238 | NP_001159857.2 |
NM_009239.4 | 5 | 2244 | NP_033265.4 | |
AB221633.1 | 4 | 2037 | ||
BC076630.1 | 5 | 2168 | AAH76630.1 | |
U62522.1 | 5 | 2173 | AAC52653.1 |
More Information
Set Membership: |
Intragenic Exonic |