Assay Details
Target Gene Details
Entrez Gene ID: | 104759 |
Gene Name: | phospholipase D family, member 4 |
Gene Aliases: |
AI132321, thss |
Location: |
Chr.12:112760655-112768988 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pld4 | NM_178911.4 | NP_849242.1 | ||
AK045459.1 | BAC32379.1 | |||
AK079549.1 | BAC37681.1 | |||
AK079878.1 | BAC37771.1 | |||
AK151551.1 | BAE30496.1 | |||
AK152969.1 | ||||
AK170362.1 | BAE41745.1 | |||
BC058565.1 | AAH58565.1 |
More Information
Set Membership: |
Intragenic Non-exonic |