Assay Details
Target Gene Details
Entrez Gene ID: | 218214 |
Gene Name: | lysine (K)-specific demethylase 1B |
Gene Aliases: |
4632428N09Rik, AI482520, Aof1 |
Location: |
Chr.13:47043471-47085279 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kdm1b | NM_172262.3 | NP_758466.1 | ||
AK028553.1 | BAC26005.1 | |||
BC023917.1 | AAH23917.1 | |||
BY714054.1 |
More Information
Set Membership: |
Intragenic Non-exonic |